Learn More
1-Methyl-2-(tri-n-butylstannyl)pyrrole, 98%, Thermo Scientific Chemicals
$122.15 - $122.15
Chemical Identifiers
| CAS | 118486-97-8 |
|---|---|
| Molecular Formula | C17H33NSn |
| Molecular Weight (g/mol) | 370.168 |
| MDL Number | MFCD01319022 |
| InChI Key | DINAKCGOEKXDTP-UHFFFAOYSA-N |
| Synonym | 1-methyl-2-tributylstannyl pyrrole, 1-methyl-2-tributylstannyl-1h-pyrrole, 1-methyl-2-tributylstannylpyrrole, 1h-pyrrole,1-methyl-2-tributylstannyl, 1-methylpyrrol-2-yl tributyltin, 1-methyl-2-tri-n-butylstannyl pyrrole, n-methyl-2-tributylstannyl-1h-pyrrole, acmc-20aotm, tributyl-1-methylpyrrol-2-yl stannane, tributylstannyl-1-methylpyrrole |
| PubChem CID | 2769397 |
| IUPAC Name | tributyl-(1-methylpyrrol-2-yl)stannane |
| SMILES | CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5143803
|
Thermo Scientific Chemicals
H5143803 |
1 g |
Each for $122.15
|
|
|||||
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 118486-97-8 | |
| 370.168 | |
| DINAKCGOEKXDTP-UHFFFAOYSA-N | |
| 2769397 | |
| CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C |
| C17H33NSn | |
| MFCD01319022 | |
| 1-methyl-2-tributylstannyl pyrrole, 1-methyl-2-tributylstannyl-1h-pyrrole, 1-methyl-2-tributylstannylpyrrole, 1h-pyrrole,1-methyl-2-tributylstannyl, 1-methylpyrrol-2-yl tributyltin, 1-methyl-2-tri-n-butylstannyl pyrrole, n-methyl-2-tributylstannyl-1h-pyrrole, acmc-20aotm, tributyl-1-methylpyrrol-2-yl stannane, tributylstannyl-1-methylpyrrole | |
| tributyl-(1-methylpyrrol-2-yl)stannane |
Specifications
| 118486-97-8 | |
| 98°C (208°F) | |
| 1.511 | |
| 1 g | |
| 1-methyl-2-tributylstannyl pyrrole, 1-methyl-2-tributylstannyl-1h-pyrrole, 1-methyl-2-tributylstannylpyrrole, 1h-pyrrole,1-methyl-2-tributylstannyl, 1-methylpyrrol-2-yl tributyltin, 1-methyl-2-tri-n-butylstannyl pyrrole, n-methyl-2-tributylstannyl-1h-pyrrole, acmc-20aotm, tributyl-1-methylpyrrol-2-yl stannane, tributylstannyl-1-methylpyrrole | |
| DINAKCGOEKXDTP-UHFFFAOYSA-N | |
| tributyl-(1-methylpyrrol-2-yl)stannane | |
| 2769397 | |
| 98% |
| 1.122 | |
| C17H33NSn | |
| MFCD01319022 | |
| UN2788 | |
| Not miscible or difficult to mix. | |
| CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C | |
| 370.168 | |
| 370.15 | |
| 1-Methyl-2-(tri-n-butylstannyl)pyrrole |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only