Learn More
1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals
$482.16 - $482.16
Chemical Identifiers
| CAS | 105494-69-7 |
|---|---|
| Molecular Formula | C16H32N2Sn |
| Molecular Weight (g/mol) | 371.156 |
| MDL Number | MFCD01319030 |
| InChI Key | KFWFYOPKNSYTMV-UHFFFAOYSA-N |
| Synonym | 1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole |
| PubChem CID | 2763252 |
| IUPAC Name | tributyl-(1-methylimidazol-2-yl)stannane |
| SMILES | CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5152303
|
Thermo Scientific Chemicals
H5152303 |
1 g |
Each for $482.16
|
|
|||||
Description
1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.
Solubility
Sparingly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
| 105494-69-7 | |
| 371.156 | |
| KFWFYOPKNSYTMV-UHFFFAOYSA-N | |
| 2763252 | |
| CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C |
| C16H32N2Sn | |
| MFCD01319030 | |
| 1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole | |
| tributyl-(1-methylimidazol-2-yl)stannane |
Specifications
| 105494-69-7 | |
| MFCD01319030 | |
| UN2788 | |
| Sparingly soluble in water. | |
| CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C | |
| 371.156 | |
| 371.15 | |
| 1-Methyl-2-(tri-n-butylstannyl)imidazole |
| C16H32N2Sn | |
| 1 g | |
| 1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole | |
| KFWFYOPKNSYTMV-UHFFFAOYSA-N | |
| tributyl-(1-methylimidazol-2-yl)stannane | |
| 2763252 | |
| ≥90% |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only