missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyridone 99.0+%, TCI America™
Supplier: TCI America M041210G
Specifications
| 1-Methyl-2-pyridone | |
| Yellow | |
| C6H7NO | |
| 10 g | |
| DVVGIUUJYPYENY-UHFFFAOYSA-N | |
| 1-methylpyridin-2-one | |
| 12755 | |
| ≥99.0% (GC) |
| 694-85-9 | |
| 252°C | |
| MFCD00006520 | |
| 1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone | |
| CN1C=CC=CC1=O | |
| 109.128 | |
| 109.13 | |
| Liquid |
Chemical Identifiers
| 694-85-9 | |
| 109.128 | |
| DVVGIUUJYPYENY-UHFFFAOYSA-N | |
| 12755 | |
| CN1C=CC=CC1=O |
| C6H7NO | |
| MFCD00006520 | |
| 1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone | |
| 1-methylpyridin-2-one |
Safety and Handling
RTECSNumber : UV1148100
TSCA : Yes