missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-piperidinemethanol 98.0+%, TCI America™

Catalog No. M092325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M092325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M092325ML Supplier TCI America Supplier No. M092325ML

Chemical Identifiers

CAS 20845-34-5
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006494
InChI Key HXXJMMLIEYAFOZ-UHFFFAOYSA-N
Synonym 1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
PubChem CID 89394
IUPAC Name (1-methylpiperidin-2-yl)methanol
SMILES CN1CCCCC1CO

Specifications

Boiling Point 98°C
Quantity 25 mL
Formula Weight 129.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-2-piperidinemethanol