missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-phenylindole 99.0+%, TCI America™

Catalog No. p-7132329
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M073325G 25 g
M0733500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M073325G Supplier TCI America Supplier No. M073325G
Only null left

Chemical Identifiers

CAS 3558-24-5
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00022892
InChI Key SFWZZSXCWQTORH-UHFFFAOYSA-N
Synonym 1-methyl-2-phenyl-1h-indole, 2-phenyl-n-methylindole, 1h-indole, 1-methyl-2-phenyl, n-methyl-2-phenylindole, indole, 1-methyl-2-phenyl, pubchem7435, n-methyl-2-phenyl indole, n-methyl-2-phenyl-indole, acmc-1cp6x, 1-methyl-2-phenyl-indole
PubChem CID 77095
IUPAC Name 1-methyl-2-phenylindole
SMILES CN1C2=CC=CC=C2C=C1C3=CC=CC=C3

Specifications

Melting Point 102°C
Color Yellow
Quantity 25 g
Formula Weight 207.28
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-2-phenylindole