missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-phenylindole, 99%

Catalog No. AAB2210509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2210509 10 g
AAB2210518 50 g
AAB2210530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2210509 Supplier Thermo Scientific Chemicals Supplier No. B2210509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3558-24-5 | C15H13N | 207.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3558-24-5
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00022892
InChI Key SFWZZSXCWQTORH-UHFFFAOYSA-N
Synonym 1-methyl-2-phenyl-1h-indole, 2-phenyl-n-methylindole, 1h-indole, 1-methyl-2-phenyl, n-methyl-2-phenylindole, indole, 1-methyl-2-phenyl, pubchem7435, n-methyl-2-phenyl indole, n-methyl-2-phenyl-indole, acmc-1cp6x, 1-methyl-2-phenyl-indole
PubChem CID 77095
IUPAC Name 1-methyl-2-phenylindole
SMILES CN1C2=CC=CC=C2C=C1C3=CC=CC=C3

Specifications

Melting Point 98°C to 99°C
Quantity 10 g
Beilstein 148770
Formula Weight 207.28
Percent Purity 99%
Chemical Name or Material 1-Methyl-2-phenylindole
EINECSNumber 222-618-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only