missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-imidazolidinone, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 304150020

Catalog No. AC304150020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Methyl-2-imidazolidinone
694-32-6
100.0
98% min. (GC)
C4H8N2O
2g
Solubility in water: .. Other solubilities: soluble in organic solvents
CN1CCNC1=O
100.12
100.12
98+%
98.0
Authentic
Glass bottle
MFCD00800601
1-methyl-2-imidazolidinone, 2-imidazolidinone, 1-methyl, 1-methyl-2-imidazolidone, n-methylimidazolidinone, acmc-209yb9, 3-methylimidazolidin-2-one, 1-methylimidazolidine-2-one, 1-methyl-imidazolidin-2-one, 2-imidazolidinone,1-methyl, 1-methyl-2-imidazolidinone #
JTPZTKBRUCILQD-UHFFFAOYSA-N
1-methylimidazolidin-2-one
567600
98+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only