missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-imidazolemethanol, 98%

Catalog No. AAH6400614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6400614 25 g
AAH6400606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6400614 Supplier Thermo Scientific Chemicals Supplier No. H6400614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17334-08-6 | C5H8N2O | 112.132 g/mol

1-Methyl-2-imidazolemethanol is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-imidazolemethanol is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid, acid chlorides, halogens, halocarbons.

Chemical Identifiers

CAS 17334-08-6
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00964673
InChI Key CDQDMLWGTVLQEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-2-yl methanol, 1-methylimidazol-2-yl methanol, 1h-imidazole-2-methanol, 1-methyl, 1-methyl-2-imidazolyl methanol, 2-hydroxymethyl-1-methylimidazole, imidazole-2-methanol, 1-methyl, 1-methyl-1h-imidazole-2-methanol, 1-methylimidazol-2-yl methan-1-ol, pubchem9047
PubChem CID 573612
IUPAC Name (1-methylimidazol-2-yl)methanol
SMILES CN1C=CN=C1CO

Specifications

Melting Point 107°C to 111°C
Quantity 25 g
Beilstein 113492
Solubility Information Slightly soluble in water.
Formula Weight 112.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Methyl-2-imidazolemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only