missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrrol-2-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC02713CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC02713CB 250 mg
CC02713DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC02713CB Supplier Thermo Scientific Chemicals Supplier No. CC02713CB

Chemical Identifiers

CAS 69807-81-4
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD02677708
InChI Key GGCBARJYVAPZJQ-UHFFFAOYSA-N
PubChem CID 2776207
IUPAC Name (1-methylpyrrol-2-yl)methanamine
SMILES CN1C=CC=C1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-pyrrol-2-yl methanamine, 1-methyl-1h-pyrrol-2-yl methylamine, 1-methylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine, 1-methyl, 1-methyl-1h-pyrrole-2-methanamine, 1-1-methyl-1h-pyrrol-2-yl methanamine, 1-1-methylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine,1-methyl, 1-methyl-1h-pyrrol-2-yl rnethylarnine, c-1-methyl-1h-pyrrol-2-yl methylamine
Formula Weight 110.16
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrrol-2-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.