missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole, 97+%

Catalog No. p-7046537
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1483106 5 g
AAL1483114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1483106 Supplier Thermo Scientific Chemicals Supplier No. L1483106
Only null left

CAS: 930-36-9 | C4H6N2 | 82.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 930-36-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00144943
InChI Key UQFQONCQIQEYPJ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255
PubChem CID 70255
ChEBI CHEBI:59025
IUPAC Name 1-methylpyrazole
SMILES CN1C=CC=N1

Specifications

Density 0.988
Boiling Point 127°C
Flash Point 35°C (95°F)
Odor Pungent
Refractive Index 1.476
Quantity 5 g
UN Number UN1993
Beilstein 105207
Formula Weight 82.11
Percent Purity ≥97%
Chemical Name or Material 1-Methyl-1H-pyrazole
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only