missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%

Catalog No. p-7036483
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54785MD 250 mg
AAH5478503 1 g
AAH5478506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54785MD Supplier Thermo Scientific Chemicals Supplier No. H54785MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 288148-34-5 | C4H5ClN2O2S | 180.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288148-34-5
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.606
MDL Number MFCD04968667
InChI Key RDAVKKQKMLINOH-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl, chloro 1-methylpyrazol-4-yl sulfone, 4-chlorosulphonyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-sulfonylchloride, 1-methyl-4-pyrazolesulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 1-methyl, 1h-pyrazole-4-sulfonylchloride,1-methyl-9ci, 4-chlorosulfonyl-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazole-4-sulphonyl chloride
PubChem CID 17024730
IUPAC Name 1-methylpyrazole-4-sulfonyl chloride
SMILES CN1C=C(C=N1)S(=O)(=O)Cl

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 179.98
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only