missing translation for 'onlineSavingsMsg'
Learn More

1-méthyl-1H-pyrazole-3-sulfonychlorure, 97%, Thermo Scientific™

Numéro de catalogue. CC48303DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC48303DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC48303DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC48303DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 89501-90-6
Formule moléculaire C4H5ClN2O2S
Poids moléculaire (g/mol) 180.606
Numéro MDL MFCD08690273
Clé InChI TWLAHGNFQBQYEL-UHFFFAOYSA-N
Synonyme 1-methyl-1h-pyrazole-3-sulfonyl chloride, 1-methyl-1h-pyrazole-3-sulfonylchloride, 1h-pyrazole-3-sulfonyl chloride, 1-methyl, 1h-pyrazole-3-sulfonylchloride, 1-methyl, pubchem14132, acmc-209zg6, n-methylpyrazole sulfonyl chloride, chloro 1-methylpyrazol-3-yl sulfone, 1-methyl-3-pyrazolesulfonyl chloride, methyl-1-h-pyrazole-3-sulfonyl chloride
PubChem CID 13474466
Nom de l’IUPAC 1-méthylpyrazole-3-chlorure de sulfonyle
SOURIRES CN1C=CC(=N1)S(=O)(=O)Cl

Spécifications

Nom ou substance chimique 1-Methyl-1H-pyrazole-3-sulfonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 180.61
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.