missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%

Catalog No. p-7077596
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456030010 1 g
AC456030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456030010 Supplier Thermo Scientific Chemicals Supplier No. 456030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 863548-52-1 | C5H9N3 | 111.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 863548-52-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
InChI Key XNTFQMKXUFFUQO-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazol-5-yl methylamine, 1h-pyrazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-pyrazole, 2-methylpyrazol-3-yl methanamine, 5-aminomethyl-1-methylpyrazole, 1-1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazole-5-methanamine, 1-methylpyrazol-5-yl methylamine, c-2-methyl-2h-pyrazol-3-yl-methylamine
PubChem CID 7019421
IUPAC Name (2-methylpyrazol-3-yl)methanamine
SMILES CN1C(=CC=N1)CN

Specifications

Boiling Point 140.0°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only