missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-3-yl)methanol, 95%, Thermo Scientific™

Catalog No. CC48309DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48309DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48309DA Supplier Thermo Scientific Chemicals Supplier No. CC48309DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84547-62-6
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD08690274
InChI Key WCKJRVKJTUIAFW-UHFFFAOYSA-N
PubChem CID 12845046
IUPAC Name (1-methylpyrazol-3-yl)methanol
SMILES CN1C=CC(=N1)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-methyl-1h-pyrazol-3-yl methanol, 1h-pyrazole-3-methanol, 1-methyl, 1-methylpyrazol-3-yl methanol, 1h-pyrazole-3-methanol,1-methyl, 3-hydroxymethyl-1-methyl-1h-pyrazole, 1-methylpyrazol-3-yl methan-1-ol, akos pao-1076, pubchem23752, 1-methyl-3-pyrazolyl methanol, hydroxymethyl-1-methylpyrazole
Formula Weight 112.13
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-pyrazol-3-yl)methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.