missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carboxylic acid, 95%, Thermo Scientific™

Catalog No. CC41401CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC41401CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC41401CB Supplier Thermo Scientific Chemicals Supplier No. CC41401CB

Chemical Identifiers

CAS 186129-25-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD03839859
InChI Key UHQAIJFIXCOBCN-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-5-indolecarboxylic acid, 1h-indole-5-carboxylic acid, 1-methyl, 1-methyl-1h-indole-5-carboxylicacid, 5-carboxy-1-methyl-1h-indole, 1h-indole-5-carboxylicacid,1-methyl, n-methylindol-5-carboxylic acid, 1-methyl-5-indolecarboxylicacid, methyl 1h-indole-5-carboxylic acid, 1-methyl-1h-indol-5-carboxylic acid
PubChem CID 7015601
IUPAC Name 1-methylindole-5-carboxylic acid
SMILES CN1C=CC2=C1C=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indole-5-carboxylic acid
Hazard Category Warning
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.