missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC41404DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC41404DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC41404DA Supplier Thermo Scientific Chemicals Supplier No. CC41404DA

Chemical Identifiers

CAS 90923-75-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08271908
InChI Key XIVDZBIBWGQOTI-UHFFFAOYSA-N
PubChem CID 7537534
SMILES CN1C=CC2=CC(C=O)=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-methyl-1h-indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 1-methyl, 5-formyl-1-methylindole, n-methylindol-5-carbaldehyde, n-methylindole-5-carbaldehyde, n-methylindole-5-carboxaldehyde, 1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde,1-methyl
IUPAC Name 1-methylindole-5-carbaldehyde
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-5-carbaldehyde
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.