missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1H-indole-4-carbonitrile, Thermo Scientific™

Catalog No. p-4391405
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45816CB 250mg
CC45816DA 1g
CC45816DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC45816CB Supplier Thermo Scientific Chemicals Supplier No. CC45816CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628711-58-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD08690265
InChI Key DYOVHBKIEBCIIE-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-4-carbonitrile, 1h-indole-4-carbonitrile,1-methyl, 1h-indole-4-carbonitrile, 1-methyl
PubChem CID 22391592
IUPAC Name 1-methylindole-4-carbonitrile
SMILES CN1C=CC2=C(C=CC=C12)C#N

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-4-carbonitrile
Hazard Category GHS Signal Word: Warning