missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-indol-5-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC41409DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC41409DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

448967-90-6
161.20
CEBLTQGXYITWTM-UHFFFAOYSA-N
24229490
CN1C=CC2=CC(CO)=CC=C12
C10H11NO
MFCD09025838
1-methyl-1h-indol-5-yl methanol, 1-methylindol-5-yl methanol, 1h-indole-5-methanol, 1-methyl-9ci
(1-methylindol-5-yl)methanol

Specifications

(1-Methyl-1H-indol-5-yl)methanol
97%
C10H11NO
5g
CEBLTQGXYITWTM-UHFFFAOYSA-N
(1-methylindol-5-yl)methanol
24229490
97%
448967-90-6
Amber Glass Bottle
MFCD09025838
1-methyl-1h-indol-5-yl methanol, 1-methylindol-5-yl methanol, 1h-indole-5-methanol, 1-methyl-9ci
CN1C=CC2=CC(CO)=CC=C12
161.20
161.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents