missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-4-carboxaldehyde, 95%

Catalog No. AC429340010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429340010 1 g Glass bottle
AC429340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429340010 Supplier Thermo Scientific Chemicals Supplier No. 429340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1053655-56-3 | C9H8N2O | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1053655-56-3
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD10699167
InChI Key SONWSIAHVBTXPB-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-4-carbaldehyde, 1-methyl-1h-indazole-4-carboxaldehyde, 4-formyl-1-methyl-1h-indazole, 1-methyl-4-indazolecarboxaldehyde
PubChem CID 37818471
IUPAC Name 1-methylindazole-4-carbaldehyde
SMILES CN1C2=CC=CC(=C2C=N1)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 77.5°C to 79.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 160.18
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1H-indazole-4-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only