missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-indazole-3-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC06302DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC06302DA 1 g Amber Glass Bottle
CC06302CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC06302DA Supplier Thermo Scientific Chemicals Supplier No. CC06302DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106649-02-9
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.62
MDL Number MFCD02093096
InChI Key NPNWVMBPSINFBV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride
PubChem CID 7022055
IUPAC Name 1-methylindazole-3-carbonyl chloride
SMILES CN1N=C(C(Cl)=O)C2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 194.62
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-3-carbonyl chloride