missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2695259
Change view
Click to view available options
Quantity:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC23601DA 1g
CC23601DE 5g
CC23601EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC23601DA Supplier Thermo Scientific Chemicals Supplier No. CC23601DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41806-40-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00955677
InChI Key PBEDVTDUVXFSMW-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carboxylic acid, 1-methylimidazole-5-carboxylic acid, 5-carboxy-1-methyl-1h-imidazole, 3-methyl-3h-imidazole-4-carboxylic acid, 1-methyl-1h-imidazole-5-carboxylicacid, 1-methyl-1h-5-imidazolecarboxylic acid, pubchem20954, acmc-209jlu, ksc236i7p, 1-methyl-5-imidazolecarboxylic acid
PubChem CID 2773507
IUPAC Name 3-methylimidazole-4-carboxylic acid
SMILES CN1C=NC=C1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.