missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carbohydrazide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 23585-00-4
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.15
MDL Number MFCD03659706
InChI Key CFIAOJVURKEWPG-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carboxylic acid hydrazide, 1h-imidazole-5-carboxylicacid, 1-methyl-, hydrazide, 1-methyl-5-imidazoliecarboxylic hydrazide, 1-methylimidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carbohydrazide #, 5-hydrazinocarbonyl-1-methyl-1h-imidazole, imidazole-1-methyl-5-carboxylic acid, hydrazide, 1h-imidazole-5-carboxylicacid,1-methyl-,hydrazide, 1h-imidazole-5-carboxylic acid, 1-methyl-, hydrazide 9ci
PubChem CID 468661
IUPAC Name 3-methylimidazole-4-carbohydrazide
SMILES CN1C=NC=C1C(=O)NN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC23627CB
SDS View Documents
Thermo Scientific Chemicals
CC23627CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC23627DA
SDS View Documents
Thermo Scientific Chemicals
CC23627DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC23627DE
SDS View Documents
Thermo Scientific Chemicals
CC23627DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23585-00-4
140.15
CFIAOJVURKEWPG-UHFFFAOYSA-N
468661
CN1C=NC=C1C(=O)NN
C5H8N4O
MFCD03659706
1-methyl-1h-imidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carboxylic acid hydrazide, 1h-imidazole-5-carboxylicacid, 1-methyl-, hydrazide, 1-methyl-5-imidazoliecarboxylic hydrazide, 1-methylimidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carbohydrazide #, 5-hydrazinocarbonyl-1-methyl-1h-imidazole, imidazole-1-methyl-5-carboxylic acid, hydrazide, 1h-imidazole-5-carboxylicacid,1-methyl-,hydrazide, 1h-imidazole-5-carboxylic acid, 1-methyl-, hydrazide 9ci
3-methylimidazole-4-carbohydrazide

Specifications

23585-00-4
100.0
Amber glass bottle
MFCD03659706
1-methyl-1h-imidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carboxylic acid hydrazide, 1h-imidazole-5-carboxylicacid, 1-methyl-, hydrazide, 1-methyl-5-imidazoliecarboxylic hydrazide, 1-methylimidazole-5-carbohydrazide, 1-methyl-1h-imidazole-5-carbohydrazide #, 5-hydrazinocarbonyl-1-methyl-1h-imidazole, imidazole-1-methyl-5-carboxylic acid, hydrazide, 1h-imidazole-5-carboxylicacid,1-methyl-,hydrazide, 1h-imidazole-5-carboxylic acid, 1-methyl-, hydrazide 9ci
CN1C=NC=C1C(=O)NN
140.15
140.15
1-Methyl-1H-imidazole-5-carbohydrazide
96.5
97%
C5H8N4O
250mg
CFIAOJVURKEWPG-UHFFFAOYSA-N
3-methylimidazole-4-carbohydrazide
468661
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'