missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™
$158.58 - $158.58
Chemical Identifiers
| CAS | 39021-62-0 |
|---|---|
| Molecular Formula | C5H6N2O |
| Molecular Weight (g/mol) | 110.116 |
| InChI Key | BNYKZFOZWZMEJD-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-imidazole-5-carbaldehyde, 1-methyl-1h-imidazole-5-carboxaldehyde, 3-methyl-3h-imidazole-4-carbaldehyde, 1-methylimidazole-5-carboxaldehyde, 1h-imidazole-5-carboxaldehyde, 1-methyl, 1-methylimidazole-5-carbaldehyde, 1-methyl-5-imidazolecarboxaldehyde, 1-methylimidazole-5-carboxaldeyde, pubchem8047, 5-formyl-1-methylimidazole |
| PubChem CID | 573592 |
| IUPAC Name | 3-methylimidazole-4-carbaldehyde |
| SMILES | CN1C=NC=C1C=O |
Chemical Identifiers
| 39021-62-0 | |
| 110.116 | |
| 1-methyl-1h-imidazole-5-carbaldehyde, 1-methyl-1h-imidazole-5-carboxaldehyde, 3-methyl-3h-imidazole-4-carbaldehyde, 1-methylimidazole-5-carboxaldehyde, 1h-imidazole-5-carboxaldehyde, 1-methyl, 1-methylimidazole-5-carbaldehyde, 1-methyl-5-imidazolecarboxaldehyde, 1-methylimidazole-5-carboxaldeyde, pubchem8047, 5-formyl-1-methylimidazole | |
| 3-methylimidazole-4-carbaldehyde |
| C5H6N2O | |
| BNYKZFOZWZMEJD-UHFFFAOYSA-N | |
| 573592 | |
| CN1C=NC=C1C=O |
Specifications
| 39021-62-0 | |
| Amber Glass Bottle | |
| 1 g | |
| BNYKZFOZWZMEJD-UHFFFAOYSA-N | |
| 3-methylimidazole-4-carbaldehyde | |
| 573592 | |
| 97% |
| 97% | |
| C5H6N2O | |
| 1-methyl-1h-imidazole-5-carbaldehyde, 1-methyl-1h-imidazole-5-carboxaldehyde, 3-methyl-3h-imidazole-4-carbaldehyde, 1-methylimidazole-5-carboxaldehyde, 1h-imidazole-5-carboxaldehyde, 1-methyl, 1-methylimidazole-5-carbaldehyde, 1-methyl-5-imidazolecarboxaldehyde, 1-methylimidazole-5-carboxaldeyde, pubchem8047, 5-formyl-1-methylimidazole | |
| CN1C=NC=C1C=O | |
| 110.116 | |
| 110.12 | |
| 1-Methyl-1H-imidazole-5-carbaldehyde |