missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC23604DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23604DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC23604DA Supplier Thermo Scientific Chemicals Supplier No. CC23604DA

Chemical Identifiers

CAS 39021-62-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
InChI Key BNYKZFOZWZMEJD-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carbaldehyde, 1-methyl-1h-imidazole-5-carboxaldehyde, 3-methyl-3h-imidazole-4-carbaldehyde, 1-methylimidazole-5-carboxaldehyde, 1h-imidazole-5-carboxaldehyde, 1-methyl, 1-methylimidazole-5-carbaldehyde, 1-methyl-5-imidazolecarboxaldehyde, 1-methylimidazole-5-carboxaldeyde, pubchem8047, 5-formyl-1-methylimidazole
PubChem CID 573592
IUPAC Name 3-methylimidazole-4-carbaldehyde
SMILES CN1C=NC=C1C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbaldehyde