missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-sulfonyl chloride, 95+%, Thermo Scientific™

Catalog No. CC03603DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03603DA 1 g Amber Glass Bottle
CC03603EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03603DA Supplier Thermo Scientific Chemicals Supplier No. CC03603DA

Chemical Identifiers

CAS 137049-00-4
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.61
MDL Number MFCD00068060
InChI Key KXUGUWTUFUWYRS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchloride
PubChem CID 2736887
IUPAC Name 1-methylimidazole-4-sulfonyl chloride
SMILES CN1C=NC(=C1)S(Cl)(=O)=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.61
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-imidazole-4-sulfonyl chloride