missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-carbonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC03616DA

Quantity 1g
Packaging Amber glass bottle

 View more versions of this product

Catalog No. CC03616DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

66121-69-5
107.116
LAKVILSENFFPRR-UHFFFAOYSA-N
2795115
CN1C=C(N=C1)C#N
C5H5N3
MFCD06411537
1-methyl-1h-imidazole-4-carbonitrile, 4-cyano-1-methyl-1h-imidazole, 1h-imidazole-4-carbonitrile, 1-methyl, 4-cyano-1-methylimidazole, 1-methyl-4-cyano-1h-imidazole, 1-methyl-4-imidazolecarbonitrile, 1h-imidazole-4-carbonitrile,1-methyl
1-methylimidazole-4-carbonitrile

Specifications

1-Methyl-1H-imidazole-4-carbonitrile
96.5
97%
C5H5N3
MFCD06411537
LAKVILSENFFPRR-UHFFFAOYSA-N
1-methylimidazole-4-carbonitrile
2795115
97%
66121-69-5
100.0
Amber glass bottle
1g
1-methyl-1h-imidazole-4-carbonitrile, 4-cyano-1-methyl-1h-imidazole, 1h-imidazole-4-carbonitrile, 1-methyl, 4-cyano-1-methylimidazole, 1-methyl-4-cyano-1h-imidazole, 1-methyl-4-imidazolecarbonitrile, 1h-imidazole-4-carbonitrile,1-methyl
CN1C=C(N=C1)C#N
107.116
107.12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions