missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-5-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC23613DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23613DA 1 g Amber Glass Bottle
CC23613CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC23613DA Supplier Thermo Scientific Chemicals Supplier No. CC23613DA

Chemical Identifiers

CAS 486414-86-2
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.148
InChI Key PYAQTQXFMQWCHQ-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-5-yl methylamine, 1-methyl-5-aminomethylimidazole, 1-methyl-1h-imidazol-5-yl methanamine, 3-methylimidazol-4-yl methanamine, 1h-imidazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-imidazole, c-3-methyl-3h-imidazol-4-yl-methylamine, 1-3-methylimidazol-4-yl methanamine, 1-methyl-imidazol-5-yl methylamine, 1-1-methyl-1h-imidazole-5-yl methanamine
PubChem CID 2795424
IUPAC Name (3-methylimidazol-4-yl)methanamine
SMILES CN1C=NC=C1CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-imidazol-5-yl)methylamine