missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-4-yl)methylamine, 95%, Thermo Scientific™

Catalog No. CC03613CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03613CB 250 mg Amber Glass Bottle
CC03613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03613CB Supplier Thermo Scientific Chemicals Supplier No. CC03613CB

Chemical Identifiers

CAS 486414-83-9
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD06411536
InChI Key YSEAGFBRAQOCFM-UHFFFAOYSA-N
PubChem CID 2795114
IUPAC Name (1-methylimidazol-4-yl)methanamine
SMILES CN1C=NC(CN)=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-imidazol-4-yl methanamine, 1-methyl-1h-imidazol-4-yl methylamine, 1h-imidazole-4-methanamine, 1-methyl, 1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methylimidazole, c-1-methyl-1h-imidazol-4-yl-methylamine, 1-1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methyl-1h-imidazole, 1h-imidazole-4-methanamine,1-methyl, 1-methyl-1h-imidazol-4-yl methyl amine
Formula Weight 111.15
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methylamine