missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride, 90%, Thermo Scientific™

Catalog No. p-3541072
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13652CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13652CB Supplier Thermo Scientific Chemicals Supplier No. CC13652CB
Only null left

Chemical Identifiers

CAS 906352-57-6
Molecular Formula C9H8Cl2N2O
Molecular Weight (g/mol) 231.08
MDL Number MFCD09817460
InChI Key SGNORNZSRSQMEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
PubChem CID 24229458
IUPAC Name 1-methylbenzimidazole-5-carbonyl chloride;hydrochloride
SMILES Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 231.08
Percent Purity 90%
Chemical Name or Material 1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.