missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2,4-triazole, 98%

Catalog No. p-7047248
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1833703 1 g
AAL1833706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1833703 Supplier Thermo Scientific Chemicals Supplier No. L1833703
Only null left

CAS: 6086-21-1 | C3H5N3 | 83.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6086-21-1
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.09
MDL Number MFCD01076192
InChI Key MWZDIEIXRBWPLG-UHFFFAOYSA-N
Synonym 1-methyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-methyl, 1-methyl,-1,2,4-triazole, 1-methyl-1h-1,2,4 triazole, pubchem22534, acmc-1avow, ksc495k1j
PubChem CID 22459
SMILES CN1C=NC=N1

Specifications

Melting Point 16°C
Density 1.1
Boiling Point 177°C to 178°C
Flash Point 81°C (177°F)
Refractive Index 1.4705
Quantity 1 g
Beilstein 106245
IUPAC Name 1-methyl-1,2,4-triazole
Formula Weight 83.1
Percent Purity 99%
Chemical Name or Material 1-Methyl-1,2,4-triazole
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 676-652-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only