missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-5-methylphenazinium Methyl Sulfate 95.0+%, TCI America™

Catalog No. M1442200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1442200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1442200MG Supplier TCI America Supplier No. M1442200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65162-13-2
Molecular Formula C15H16N2O5S
Molecular Weight (g/mol) 336.362
MDL Number MFCD00040648
InChI Key MASUWVVNWALEEM-UHFFFAOYSA-M
PubChem CID 127832
IUPAC Name 1-methoxy-5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=C(C2=NC3=CC=CC=C31)OC.COS(=O)(=O)[O-]

Specifications

Melting Point 172°C
Quantity 200 mg
Formula Weight 336.36
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methoxy-5-methylphenazinium Methyl Sulfate [for Biochemical Research]