missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-4-methylnaphthalene 97.0+%, TCI America™

Catalog No. M2930200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2930200MG 200 mg
M29301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2930200MG Supplier TCI America Supplier No. M2930200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24821-54-3
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.23
MDL Number MFCD00021472
InChI Key DUXRFNPXLWUXMT-UHFFFAOYSA-N
PubChem CID 4381467
IUPAC Name 1-methoxy-4-methylnaphthalene
SMILES COC1=C2C=CC=CC2=C(C)C=C1

Specifications

Boiling Point 155°C
Quantity 200 mg
Formula Weight 172.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-4-methylnaphthalene