missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methoxy-3-trimethylsiloxy-1,3-butadiene, 96%
Description
1-Methoxy-3-trimethylsiloxy-1,3-butadiene is employed as a reagent in Mannich-Michael reaction for the synthesis of piperidinones and enaminones. As Diels-Alder diene, it is used in the synthesis of pyridones and pyranones, sulfone analogues of griseofulvin (sulfogriseofulvins) and 4H-1-aminopyrroles and 4, 5H-pyrazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methoxy-3-trimethylsiloxy-1,3-butadiene is employed as a reagent in Mannich-Michael reaction for the synthesis of piperidinones and enaminones. As Diels-Alder diene, it is used in the synthesis of pyridones and pyranones, sulfone analogues of griseofulvin (sulfogriseofulvins) and 4H-1-aminopyrroles and 4, 5H-pyrazoles.
Solubility
Miscible with most organic solvents. Immiscible with water.
Notes
Moisture sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
1-Methoxy-3-trimethylsiloxy-1,3-butadiene is employed as a reagent in Mannich-Michael reaction for the synthesis of piperidinones and enaminones. As Diels-Alder diene, it is used in the synthesis of pyridones and pyranones, sulfone analogues of griseofulvin (sulfogriseofulvins) and 4H-1-aminopyrroles and 4, 5H-pyrazoles.
Solubility
Miscible with most organic solvents. Immiscible with water.
Notes
Moisture sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Boiling Point | 68°C to 69°C (14mmHg) |
| Quantity | 5 g |
| Flash Point | 45°C (113°F) |
| Refractive Index | 1.455 |
| Sensitivity | Air and moisture sensitive |
| Density | 0.89 |
| UN Number | UN1993 |
| Beilstein | 1616761 |
| Merck Index | 14,6001 |
| Solubility Information | Miscible with most organic solvents. Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only