missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol 98.0+%, TCI America™

Catalog No. p-7132007
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012625ML 25 mL
M0126500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M012625ML Supplier TCI America Supplier No. M012625ML
Only null left

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Melting Point -96°C
Color Colorless
Boiling Point 120°C
Quantity 25 mL
UN Number 3092
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
EINECSNumber (2)-0404
RTECSNumber UB7700000
TSCA Yes