missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol Acetate, 97%, Spectrum™ Chemical

Catalog No. p-7157777
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600054 1 L Amber Glass Bottle
18600055 4 L Amber Glass Bottle
18600056 20 L Steel Can
18600057 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600054 Supplier Spectrum Chemical Mfg Cor Supplier No. M26341LT
Only null left

M2634, 108-65-6, C6H12O3

Spectrum™ Chemical 1-Methoxy-2-propanol Acetate

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
InChI Key LLHKCFNBLRBOGN-UHFFFAOYNA-N
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES COCC(C)OC(C)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 L
UN Number UN3272
Percent Purity 97%
Grade Reagent