missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 99+%

Catalog No. AA41547M6
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41547M6 4 x 4 L
AA41547AK 250 mL
AA41547K2 1 L
AA41547K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA41547M6 Supplier Thermo Scientific Chemicals Supplier No. 041547M6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Melting Point -100°C
Color Colorless
Density 0.926 g/mL
pH 4 to 7
Boiling Point 119°C to 120°C
Flash Point 35°C (95°F)
Odor Alcohol-like
Assay Percent Range >99%
Refractive Index 1.403
Quantity 4 x 4 L
UN Number UN3092
Beilstein 1731270
Solubility Information Miscible with water,methanol and ether.
Formula Weight 90.12
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
Show More Show Less
Hazard Category H226-H335-H336
Hazard Statement GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-METHOXY-2-PROPANOL
EINECSNumber 203-539-1
RTECSNumber UB7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only