missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 98.5%, extra pure

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

Supplier:  Thermo Scientific Chemicals 244990010

 View more versions of this product

Catalog No. AC244990010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methoxy-2-propanol
-97.0°C
120.0°C
Authentic
Glass Bottle
1.4020 to 1.4040
MFCD00004537
01,536
0.92
Solubility in water: 1000g/L (20°C)
COCC(C)O
90.12
1.7 mPa.s (20°C)
98.5%
Liquid
107-98-2
0.9200g/mL
30°C
98% min. (GC)
C4H10O2
CH3OCH2CH(OH)CH3
1 L
0.01% max.
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
ARXJGSRGQADJSQ-UHFFFAOYNA-N
1-methoxypropan-2-ol
7900
90.12
Extra Pure

Chemical Identifiers

107-98-2
90.12
ARXJGSRGQADJSQ-UHFFFAOYNA-N
7900
COCC(C)O
C4H10O2
MFCD00004537
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
1-methoxypropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Danger

EINECSNumber : 203-539-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only