missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 98.5%, extra pure

Catalog No. AC244990250
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC244990250 25 mL Glass Bottle
AC244990010 1 L Glass Bottle
AC244990025 2.5 L Glass Bottle
AC244990050 5 L Plastic drum
AC244990251 25 L Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC244990250 Supplier Thermo Scientific Chemicals Supplier No. 244990250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Melting Point -97.0°C
Density 0.9200g/mL
Boiling Point 120.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4020 to 1.4040
Linear Formula CH3OCH2CH(OH)CH3
Quantity 25 mL
Beilstein 01,536
Residue after Evaporation 0.01% max.
Specific Gravity 0.92
Solubility Information Solubility in water: 1000g/L (20°C)
Viscosity 1.7 mPa.s (20°C)
Formula Weight 90.12
Percent Purity 98.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
EINECSNumber 203-539-1

RUO – Research Use Only