missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-methyl-2-propanol, 98+%

Catalog No. AAL2035122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2035122 100 g
AAL2035114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2035122 Supplier Thermo Scientific Chemicals Supplier No. L2035122

CAS: 3587-64-2 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3587-64-2
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD03701584
InChI Key MXUXZWFVAPTPAG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol #
PubChem CID 77137
IUPAC Name 1-methoxy-2-methylpropan-2-ol
SMILES COCC(C)(C)O

Specifications

Density 0.892
Boiling Point 115°C to 116°C
Flash Point 27°C (80°F)
Refractive Index 1.4044
Quantity 100 g
UN Number UN1987
Formula Weight 104.15
Percent Purity ≥98%
Chemical Name or Material 1-Methoxy-2-methyl-2-propanol
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only