missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-butanol 93.0+%, TCI America™

Catalog No. M074725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M074725ML 25 mL
M0747500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M074725ML Supplier TCI America Supplier No. M074725ML
Only null left

Chemical Identifiers

CAS 53778-73-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00059143
InChI Key CSZZMFWKAQEMPB-UHFFFAOYSA-N
Synonym 1,2-Butanediol 1-Monomethyl Ether
PubChem CID 40895
IUPAC Name 1-methoxybutan-2-ol
SMILES CCC(COC)O

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.15
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-butanol
RTECSNumber EL4540000
TSCA Yes