missing translation for 'onlineSavingsMsg'
Learn More

1-Kestose 99.0+%, TCI America™

Catalog No. K00325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00325G 5 g
K00321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00325G Supplier TCI America Supplier No. K00325G

Chemical Identifiers

CAS 470-69-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00142647
InChI Key VAWYEUIPHLMNNF-OESPXIITSA-N
PubChem CID 440080
ChEBI CHEBI:16885
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-2-[[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)COC3(C(C(C(O3)CO)O)O)CO)O)O)O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 504.44
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Kestose