missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences 1% Janus Green, Aqueous
Description
The indicator Janus Green changes colour according to the amount of oxygen present. When oxygen is present, the indicator oxidizes to a blue colour. In the absence of oxygen, the indicator is reduced and changes to a pink colour.
Specifications
Specifications
| Chemical Name or Material | Janus Green |
| CAS | 2869-83-2 |
| Molecular Weight (g/mol) | 511.06 g/mol |
| Percent Purity | >65% |
| Grade | Research |
| Color | Green |
| Quantity | 50 mL |