Learn More
1-Isoindolinone-5-boronic acid pinacol ester, 96%
CAS: 376584-62-2 | C14H18BNO3 | 259.112 g/mol
Supplier: Thermo Scientific Chemicals H5231703
Description
1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 1-Isoindolinone-5-boronic acid pinacol ester | |
| C14H18BNO3 | |
| 1 g | |
| Slightly soluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C(=O)NC3 | |
| 259.112 | |
| 259.11 |
| 376584-62-2 | |
| MFCD11040393 | |
| 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoindolin-1-one, 1-isoindolinone-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-isoindolinone, 1-oxoisoindoline-5-boronic acid pinacol ester, 2,3-dihydro-1h-isoindol-1-one-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-isoindol-1-one | |
| CLACMCLRELMFLJ-UHFFFAOYSA-N | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroisoindol-1-one | |
| 21955795 | |
| 96% |
Chemical Identifiers
| 376584-62-2 | |
| 259.112 | |
| CLACMCLRELMFLJ-UHFFFAOYSA-N | |
| 21955795 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C(=O)NC3 |
| C14H18BNO3 | |
| MFCD11040393 | |
| 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoindolin-1-one, 1-isoindolinone-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-isoindolinone, 1-oxoisoindoline-5-boronic acid pinacol ester, 2,3-dihydro-1h-isoindol-1-one-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-isoindol-1-one | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroisoindol-1-one |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only