missing translation for 'onlineSavingsMsg'
Learn More

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 827614-66-4
Molecular Formula C13H23BN2O2
Molecular Weight (g/mol) 250.15
InChI Key YMEBZRNYQBODKB-UHFFFAOYSA-N
Synonym 1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
PubChem CID 2759392
IUPAC Name 1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439730010
SDS View Documents
Thermo Scientific Chemicals
439730010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439730050
SDS View Documents
Thermo Scientific Chemicals
439730050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

827614-66-4
250.15
1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
C13H23BN2O2
YMEBZRNYQBODKB-UHFFFAOYSA-N
2759392
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC(C)C

Specifications

827614-66-4
100.0
248°C to 249°C
Authentic
Glass Bottle
1g
1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC(C)C
250.15
250.15
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
96.0
1.0030g/mL
48°C
96% min. (GC)
C13H23BN2O2
1.003
YMEBZRNYQBODKB-UHFFFAOYSA-N
1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
2759392
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair):

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only