missing translation for 'onlineSavingsMsg'
Learn More

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

Catalog No. AC439730050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439730050 5 g
AC439730010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439730050 Supplier Thermo Scientific Chemicals Supplier No. 439730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827614-66-4 | C13H23BN2O2 | 250.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 827614-66-4
Molecular Formula C13H23BN2O2
Molecular Weight (g/mol) 250.15
MDL Number MFCD05663857
InChI Key YMEBZRNYQBODKB-UHFFFAOYSA-N
Synonym 1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
PubChem CID 2759392
IUPAC Name 1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0030g/mL
Boiling Point 248°C to 249°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.003
Formula Weight 250.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair):

RUO – Research Use Only