missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7131192
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006825G 25 g
I0068100G 100 g
I0068500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I006825G Supplier TCI America Supplier No. I006825G
Only null left

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
UN Number 2392
Formula Weight 169.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodopropane (stabilized with Copper chip)
RTECSNumber TZ4100000
TSCA Yes