missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 98+%, stab. with copper

Catalog No. AAA1438822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1438822 100 g
AAA1438836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1438822 Supplier Thermo Scientific Chemicals Supplier No. A1438822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-08-4 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
SMILES CC(C)I

Specifications

Melting Point °C to 101°C
Density 1.744
Boiling Point 101°C to 102°C
Flash Point 44°C (111°F)
Assay Percent Range ≥98%, Stabilized with copper
Refractive Index 1.505
Linear Formula CH3CH2CH2I
Quantity 100 g
UN Number UN2392
Beilstein 505937
Sensitivity Light sensitive
Merck Index 14,7862
Solubility Information Fully miscible in water.
IUPAC Name 1-iodopropane
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, stabilized with copper
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: IODOPROPANES
EINECSNumber 203-460-2
RTECSNumber TZ4100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only