missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctane, 98%, stabilized

Catalog No. AC412465000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412465000 500 g Glass bottle
AC412461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412465000 Supplier Thermo Scientific Chemicals Supplier No. 412465000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-27-6 | C8H17I | 240.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.13
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -46°C to 45°C
Color Colorless to Amber
Density 1.3300g/mL
Boiling Point 225.0°C to 226.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4878
Linear Formula CH3(CH2)7I
Quantity 500 g
Beilstein 01,16
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Stabilizer copper
Formula Weight 240.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodooctane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-084-5

RUO – Research Use Only