missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctadecane 98.0+%, TCI America™

Catalog No. I006525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006525G 25 g
I0065100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I006525G Supplier TCI America Supplier No. I006525G
Only null left

Chemical Identifiers

CAS 629-93-6
Molecular Formula C18H37I
Molecular Weight (g/mol) 380.398
MDL Number MFCD00001090
InChI Key ZNJOCVLVYVOUGB-UHFFFAOYSA-N
Synonym Octadecyl Iodide, Stearyl Iodide
PubChem CID 12402
IUPAC Name 1-iodooctadecane
SMILES CCCCCCCCCCCCCCCCCCI

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 380.40
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1-Iodooctadecane