missing translation for 'onlineSavingsMsg'
Learn More

1-Iodononane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I049325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I049325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I049325G Supplier TCI America Supplier No. I049325G
Only null left

Chemical Identifiers

CAS 4282-42-2
Molecular Formula C9H19I
Molecular Weight (g/mol) 254.155
MDL Number MFCD00001107
InChI Key OGSJMFCWOUHXHN-UHFFFAOYSA-N
Synonym nonyl iodide, n-nonyl iodide, nonane, 1-iodo, 1-n-nonyl iodide, iodononane, nonane,1-iodo, 1-iodanylnonane, 1-iodo-n-nonane, nonane, iodo, pubchem3958
PubChem CID 20275
IUPAC Name 1-iodononane
SMILES CCCCCCCCCI

Specifications

Boiling Point 122°C
Quantity 25 g
Formula Weight 254.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodononane (stabilized with Copper chip)